Learn More
Biomatik Corporation Human IL-12 (p35) (Interleukin 12 p35) ELISA Kit
Supplier:  Biomatik Corporation EKF58599
Encompass_Preferred
Human IL-12 (p35) (Interleukin 12 p35) ELISA Kit for detecting IL-12(p35) in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P29459. Target Synonyms: Interleukin 12 p35. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5897