Learn More
Biomatik Corporation Human TM (Thrombomodulin) ELISA Kit
Supplier:  Biomatik Corporation EKF58607
Encompass_Preferred
Human TM (Thrombomodulin) ELISA Kit for detecting TM in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000pg/ml. Sensitivity: 37.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P07204. Target Synonyms: TM; Thrombomodulin; THBD; AHUS6; BDCA3; CD141; THPH12; THRM; BDCA-3; Fetomodulin; CD141 antigen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5903