Learn More
Biomatik Corporation Human CNDP1 (Beta-Ala-His dipeptidase) ELISA Kit
Supplier:  Biomatik Corporation EKF58612
Encompass_Preferred
Human CNDP1 (Beta-Ala-His dipeptidase) ELISA Kit for detecting CNDP1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.563-100ng/ml. Sensitivity: 0.938ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q96KN2. Target Synonyms: CNDP1; Carnosine Dipeptidase 1; CPGL2; CN1; Carnosinase 1; beta-Ala-His dipeptidase; carnosine dipeptidase 1; metallopeptidase M20 family; CNDP dipeptidase 1; Glutamate carboxypeptidase-like protein 2; Serum carnosinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5908