Learn More
Biomatik Corporation Mouse Gusb (Beta-glucuronidase) ELISA Kit
Supplier:  Biomatik Corporation EKF58621
Encompass_Preferred
Mouse Gusb (Beta-glucuronidase) ELISA Kit for detecting Gusb in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.813-500pg/ml. Sensitivity: 4.688pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P12265. Target Synonyms: Gusb; Beta-G1; MPS7; beta-D-glucuronidase; beta-G1; Beta-G1; BG; EC 3.2.1.31; FLJ39445; glucuronidase; beta; MPS7. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5914