Learn More
Biomatik Corporation Rat Slc6a4 (Sodium-dependent serotonin transporter) ELISA Kit
Supplier:  Biomatik Corporation EKF58637
Encompass_Preferred
Rat Slc6a4 (Sodium-dependent serotonin transporter) ELISA Kit for detecting Slc6a4 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P31652. Target Synonyms: Slc6a4; 5HT transporter; 5HTT; Solute carrier family 6 member 4;. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5927