Learn More
Biomatik Corporation Human CA1 (Carbonic anhydrase 1) ELISA Kit
Supplier:  Biomatik Corporation EKF58641
Encompass_Preferred
Human CA1 (Carbonic anhydrase 1) ELISA Kit for detecting CA1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P00915. Target Synonyms: CA1; Carbonic Anhydrase I; CA-I; Car1; Carbonate dehydRatase I; Carbonic anhydrase B; carbonic anhydrase ICAB; carbonic dehydRatase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5930