Learn More
Biomatik Corporation Human CDT1 (DNA replication factor Cdt1) ELISA Kit
Supplier:  Biomatik Corporation EKF58660
Encompass_Preferred
Human CDT1 (DNA replication factor Cdt1) ELISA Kit for detecting CDT1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9H211. Target Synonyms: CDT1; DNA replication factor Cdt1; Double parked homolog; DUP; RIS2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5945