Learn More
Biomatik Corporation Human MAP1LC3A (Microtubule-associated proteins 1A/1B light chain 3A) ELISA Kit
Supplier:  Biomatik Corporation EKF58676
Encompass_Preferred
Human MAP1LC3A (Microtubule-associated proteins 1A/1B light chain 3A) ELISA Kit for detecting MAP1LC3A in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 7.813-500pg/ml. Sensitivity: 4.688pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9H492. Target Synonyms: MAP1LC3A; Apg8; APG8a; Apg8p3; ATG8E; MAP1ALC3; MLP3A; ATG8E; Autophagy-related protein LC3 A; Autophagy-related ubiquitin-like modifier LC3 A; LC3; LC3A; lc3-i; ii; MAP1A; 1B light chain 3 A; MAP1ALC3MAP1A; MAP1B LC3 A; MAP1BLC3MAP1A; MAP1B light chain 3 A; microtubule-associated protein 1 light chain 3 alphaMAP1 light chain 3-like protein 1; microtubule-associated proteins 1A; 1B light chain 3; microtubule-associated proteins 1A; 1B light chain 3A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5957