Learn More
Biomatik Corporation Rat Tk1 (Thymidine kinase, cytosolic) ELISA Kit
Supplier:  Biomatik Corporation EKF58686
Encompass_Preferred
Rat Tk1 (Thymidine kinase, cytosolic) ELISA Kit for detecting Tk1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P27158. Target Synonyms: Tk1; thymidine kinase 1 soluble isoform; thymidine kinase; cytosolic; thymidine kinase-1; TK2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5965