Learn More
Biomatik Corporation Human KIF5A (Kinesin heavy chain isoform 5A) ELISA Kit
Supplier:  Biomatik Corporation EKF58696
Encompass_Preferred
Human KIF5A (Kinesin heavy chain isoform 5A) ELISA Kit for detecting KIF5A in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q12840. Target Synonyms: KIF5A; NKHC; MY050; SPG10; Kinesin heavy chain neuron-specific 1; Neuronal kinesin heavy chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5972