Learn More
Biomatik Corporation Human SPARCL1 (SPARC-like protein 1) ELISA Kit
Supplier:  Biomatik Corporation EKF58709
Encompass_Preferred
Human SPARCL1 (SPARC-like protein 1) ELISA Kit for detecting SPARCL1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q14515. Target Synonyms: SPARCL1; SPARC Like Protein 1; Hevin; MAST9; SC1; Hevin; High endothelial venule protein; MAST 9; mast9; PIG33; prolifeRation-inducing protein 33; SC1; SPARC-like 1; mast9; hevin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5983