Learn More
Biomatik Corporation Rat ANG-1 (Angiopoietin-1) ELISA Kit
Supplier:  Biomatik Corporation EKF58724
Encompass_Preferred
Rat ANG-1 (Angiopoietin-1) ELISA Kit for detecting ANG-1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O35460. Target Synonyms: ANG1; Angiopoietin 1; ANG-1; ANGPT1; AGP1; AGPT. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5998