Learn More
Biomatik Corporation Mouse MAPτ (Microtubule Associated Protein Tau/Tau Protein) ELISA Kit
Supplier:  Biomatik Corporation EKF58731
Encompass_Preferred
Mouse MAPτ (Microtubule Associated Protein Tau/Tau Protein) ELISA Kit for detecting MAPτ in serum, plasma, cell culture supernatant, cell or tissue lysate, cerebrospinal fluid (csf), other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P10637. Target Synonyms: MAPτ; Neurofibrillary tangle protein; Paired helical filament-tau; PHF-tau; Mtapt; Tau; Mapt. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6004