Learn More
Biomatik Corporation Human MIP-3α/CCL20 (Macrophage Inflammatory Protein 3 Alpha) ELISA Kit
Supplier:  Biomatik Corporation EKF58733
Encompass_Preferred
Human MIP-3α/CCL20 (Macrophage Inflammatory Protein 3 Alpha) ELISA Kit for detecting MIP-3α; CCL20 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P78556. Target Synonyms: MIP-3α; MIP-3a; CCL20; CKb4; LARC; MIP3A; SCYA20; ST38. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6006