Learn More
Biomatik Corporation Human CLDN4 (Claudin 4) ELISA Kit
Supplier:  Biomatik Corporation EKF58738
Encompass_Preferred
Human CLDN4 (Claudin 4) ELISA Kit for detecting CLDN4 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O14493. Target Synonyms: CLDN4; CLDN4; CPE-R; CPER; CPETR; CPETR1; WBSCR8; hCPE-R; Clostridium perfringens enterotoxin receptor; CPE-R; CPE-receptor; Williams-Beuren syndrome chromosomal region 8 protein; Clostridium perfringens enterotoxin receptor; CPE-R; CPE-receptor; Williams-Beuren syndrome chromosomal region 8 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6009