Learn More
Biomatik Corporation Rat Chat (Choline O-acetyltransferase) ELISA Kit
Supplier:  Biomatik Corporation EKF58743
Encompass_Preferred
Rat Chat (Choline O-acetyltransferase) ELISA Kit for detecting Chat in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P32738. Target Synonyms: Chat; Choline O-acetyltransferase; CHOACTase; ChAT; Choline acetylase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6014