Learn More
Biomatik Corporation Rat TRAIL/TNFSF10 (Tumor Necrosis Factor Related Apoptosis Inducing Ligand) ELISA Kit
Supplier:  Biomatik Corporation EKF58747
Encompass_Preferred
Rat TRAIL/TNFSF10 (Tumor Necrosis Factor Related Apoptosis Inducing Ligand) ELISA Kit for detecting TRAIL; TNFSF10 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q8K3G0. Target Synonyms: TRAIL; TNFSF10; CD253; Apo-2 ligand; APO2L; Apo-2L; tumor necrosis factor; ligand family; member 10. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6018