Learn More
Biomatik Corporation Mouse Vasn (Vasorin) ELISA Kit
Supplier:  Biomatik Corporation EKF58760
Encompass_Preferred
Mouse Vasn (Vasorin) ELISA Kit for detecting Vasn in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9CZT5. Target Synonyms: Vasn; Protein slit-like 2; Atia; Slitl2; VASN; Protein slit-like 2; SLITL2; slit-like 2; slit-like 2; Drosophila; vasorin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6030