Learn More
Biomatik Corporation Human TPT1 (Translationally-controlled tumor protein) ELISA Kit
Supplier:  Biomatik Corporation EKF58763
Encompass_Preferred
Human TPT1 (Translationally-controlled tumor protein) ELISA Kit for detecting TPT1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P13693. Target Synonyms: TPT1; Translationally-controlled Tumor Protein 1; TCTP; Fortilin; HRF; Histamine-releasing factor; p23; translationally-controlled tumor protein; tumor protein; translationally-controlled 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6032