Learn More
Biomatik Corporation Human UCMA (Unique cartilage matrix-associated protein) ELISA Kit
Supplier:  Biomatik Corporation EKF58765
Encompass_Preferred
Human UCMA (Unique cartilage matrix-associated protein) ELISA Kit for detecting UCMA in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q8WVF2. Target Synonyms: Gla-rich protein; C10orf49; GRP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6034