Learn More
Biomatik Corporation Human MCL1 (Induced myeloid leukemia cell differentiation protein Mcl-1) ELISA Kit
Supplier:  Biomatik Corporation EKF58770
Encompass_Preferred
Human MCL1 (Induced myeloid leukemia cell differentiation protein Mcl-1) ELISA Kit for detecting MCL1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q07820. Target Synonyms: MCL1; BCL2L3; bcl2-L-3; Bcl2-L-3; MGC104264; Bcl-2-like protein 3; Bcl-2-related protein EAT; mcl1; EAT; induced myeloid leukemia cell differentiation protein Mcl-1; Mcl-1; mcl1; EAT; MCL1-ES; MCL1L; MCL1S; MGC1839; myeloid cell leukemia ES; myeloid cell leukemia sequence 1; BCL2-related; TM. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6038