Learn More
Biomatik Corporation Rat BCL-2 (B-Cell Leukemia/Lymphoma 2) ELISA Kit
Supplier:  Biomatik Corporation EKF58774
Encompass_Preferred
Rat BCL-2 (B-Cell Leukemia/Lymphoma 2) ELISA Kit for detecting BCL-2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P49950. Target Synonyms: BCL-2; B-Cell Leukemia; Lymphoma 2; apoptosis regulator Bcl-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6042