Learn More
Biomatik Corporation Rat ES (Endostatin) ELISA Kit
Supplier:  Biomatik Corporation EKF58797
Encompass_Preferred
Rat ES (Endostatin) ELISA Kit for detecting ES in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: F1LR02. Target Synonyms: COL18A1; antiangiogenic agent; collagen alpha-1; XVIII chain; collagen; type XVIII; alpha 1; Human type XVIII collagen10endostatin; KNO; KNO1MGC74745; Knobloch syndrome; type 1; KS; multi-functional protein MFP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6062