Learn More
Biomatik Corporation Human CEBPZ (CCAAT/enhancer binding protein zeta) ELISA Kit
Supplier:  Biomatik Corporation EKF58808
Encompass_Preferred
Human CEBPZ (CCAAT/enhancer binding protein zeta) ELISA Kit for detecting CEBPZ in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q03701. Target Synonyms: CCAAT-box-binding transcription factor; CBF; CCAAT-binding factor; CBF2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6071