Learn More
Biomatik Corporation Human HAS2 (Hyaluronan synthase 2) ELISA Kit
Supplier:  Biomatik Corporation EKF58811
Encompass_Preferred
Human HAS2 (Hyaluronan synthase 2) ELISA Kit for detecting HAS2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q92819. Target Synonyms: HAS2; Hyaluronan synthase 2; HA synthase 2; Hyaluronate synthase 2; Hyaluronic acid synthase 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6074