Learn More
Biomatik Corporation Mouse Enpp2 (Ectonucleotide pyrophosphatase/phosphodiesterase family member 2) ELISA Kit
Supplier:  Biomatik Corporation EKF58816
Encompass_Preferred
Mouse Enpp2 (Ectonucleotide pyrophosphatase/phosphodiesterase family member 2) ELISA Kit for detecting Enpp2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9R1E6. Target Synonyms: Enpp2; ENPP2; E-NPP 2; Autotaxin; ATX; Autotaxin; Lysophosphatidic Acid; NPP2; PDNP2; ATX-X; AUTOTAXIN; autotaxin-t; ectonucleotide pyrophosphatase; phosphodiesterase 2; Extracellular lysophospholipase D; LysoPLD; PD-IALPHA; phosphodiesterase I; nucleotide pyrophosphatase 2; plasma lysophospholipase D. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6077