Learn More
Biomatik Corporation Human MUC17 (Mucin 17) ELISA Kit
Supplier:  Biomatik Corporation EKF58818
Encompass_Preferred
Human MUC17 (Mucin 17) ELISA Kit for detecting MUC17 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q685J3. Target Synonyms: MUC17; Mucin 17. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6079