Learn More
Biomatik Corporation Rat LPO (Lipid Peroxide) ELISA Kit
Supplier:  Biomatik Corporation EKF58825
Encompass_Preferred
Rat LPO (Lipid Peroxide) ELISA Kit for detecting LPO in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 3.125-200ng/ml. Sensitivity: 1.875ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: D4A400. Target Synonyms: LPO; Lactoperoxidase; Salivary peroxidase; SPO. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6084