Learn More
Biomatik Corporation Human BAX (Apoptosis regulator BAX) ELISA Kit
Supplier:  Biomatik Corporation EKF58836
Encompass_Preferred
Human BAX (Apoptosis regulator BAX) ELISA Kit for detecting BAX in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.781-50ng/ml. Sensitivity: 0.469ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q07812. Target Synonyms: BAX; apoptosis regulator BAX; BCL2-associated X protein; Bcl2-L-4; BCL2L4bcl2-L-4; Bcl-2-like protein 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6094