Learn More
Biomatik Corporation Human BDH1 (D-beta-hydroxybutyRate dehydrogenase, mitochondrial) ELISA Kit
Supplier:  Biomatik Corporation EKF58859
Encompass_Preferred
Human BDH1 (D-beta-hydroxybutyRate dehydrogenase, mitochondrial) ELISA Kit for detecting BDH1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q02338. Target Synonyms: BDH1; D-beta-hydroxybutyRate dehydrogenase; mitochondrial; BDH; 3-hydroxybutyRate dehydrogenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6112