Learn More
Biomatik Corporation Human CX3CR1 (CX3C chemokine receptor 1) ELISA Kit
Supplier:  Biomatik Corporation EKF58863
Encompass_Preferred
Human CX3CR1 (CX3C chemokine receptor 1) ELISA Kit for detecting CX3CR1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P49238. Target Synonyms: CX3CR1; CMKbRL1; V28; Beta chemokine receptor-like 1; chemokine; C-C receptor-like 1; CMKBRL1; CMK-BRL1; CMK-BRL-1; CMKDR1; CX3C chemokine receptor 1; Fractalkine receptor; G protein-coupled receptor 13; GPR13G-protein coupled receptor 13; GPRV28; V28C-X3-C CKR-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6116