Learn More
Biomatik Corporation Rat AgRP (Agouti Related Protein) ELISA Kit
Supplier:  Biomatik Corporation EKF58870
Encompass_Preferred
Rat AgRP (Agouti Related Protein) ELISA Kit for detecting AgRP in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9QXJ3. Target Synonyms: Agrp; AGRP; Agouti-related protein; AGRT; ASIP2; ART; agouti-related protein; Agrt. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6120