Learn More
Biomatik Corporation Human Interferon Epsilon (Ifne) Elisa Kit
Supplier:  Biomatik Corporation EKC34197
Encompass_Preferred
Human Interferon Epsilon (Ifne) Elisa Kit for detecting Interferon, Epsilon in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q86WN2; IFNE. Target Synonyms: IFNE; IFNE1; UNQ360/PRO655; Interferon epsilon; IFN-epsilon; Interferon epsilon-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4063