Learn More
Biomatik Corporation Human Interleukin-15 Receptor Subunit Alpha (Il15Ra) Elisa Kit
Supplier:  Biomatik Corporation EKC34271
Encompass_Preferred
Human Interleukin-15 Receptor Subunit Alpha (Il15Ra) Elisa Kit for detecting Interleukin 15 Receptor, Alpha in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q13261; IL15RA. Target Synonyms: AA690181; CD215; I15RA_HUMAN; IL 15R alpha; IL-15 receptor subunit alpha; IL-15R-alpha; IL-15RA; Il15ra; Interleukin 15 receptor alpha; Interleukin 15 receptor subunit alpha; MGC104179; sIL-15 receptor subunit alpha; sIL-15R-alpha; sIL-15RA; Soluble interleukin 15 receptor subunit alpha; Soluble interleukin-15 receptor subunit alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4070