Learn More
Biomatik Corporation Human Ketohexokinase (Khk) Elisa Kit
Supplier:  Biomatik Corporation EKC34346
Encompass_Preferred
Human Ketohexokinase (Khk) Elisa Kit for detecting Ketohexokinase (Fructokinase) in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 18.75-1200mU/ml. Sensitivity: 4.68mU/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P50053; KHK. Target Synonyms: EC 2.7.1.3; Fructokinase; Hepatic fructokinase; Ketohexokinase; Ketohexokinase isoform a; KHK; KHK_HUMAN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4083