Learn More
Biomatik Corporation Human Lactase-Phlorizin Hydrolase (Lct) Elisa Kit
Supplier:  Biomatik Corporation EKC34360
Encompass_Preferred
Human Lactase-Phlorizin Hydrolase (Lct) Elisa Kit for detecting Lactase in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 34.38-2200pg/ml. Sensitivity: 8.59pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P09848; LCT. Target Synonyms: LAC; Lactase; Lactase phlorizin hydrolase 1; Lactase phlorizin hydrolase; Lactase-glycosylceramidase; Lct; LPH; LPH_HUMAN; LPH1; Phlorizin hydrolase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4086