Learn More
Biomatik Corporation Human Laminin Subunit Alpha-3 (Lama3) Elisa Kit
Supplier:  Biomatik Corporation EKC34368
Encompass_Preferred
Human Laminin Subunit Alpha-3 (Lama3) Elisa Kit for detecting Laminin, Alpha 3 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 9.4-600ng/ml. Sensitivity: 2.35ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q16787; LAMA3. Target Synonyms: E170; Epiligrin 170 kDa subunit; Epiligrin subunit alpha; Kalinin B1 chain; Kalinin subunit alpha; LAMA3; LAMA3_HUMAN; LAMB3; LAMC2; Laminin gamma 2; Laminin 5 beta 3; Laminin 5 gamma 2 subunit; Laminin beta 3; Laminin subunit alpha-3; Laminin subunit beta 3; Laminin subunit gamma 2; Laminin-5 subunit alpha; Laminin-6 subunit alpha; Laminin-7 subunit alpha; Nicein subunit alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4088