Learn More
Biomatik Corporation Human Latent-Transforming Growth Factor Beta-Binding Protein 1 (Ltbp1) Elisa Kit
Supplier:  Biomatik Corporation EKC34377
Encompass_Preferred
Human Latent-Transforming Growth Factor Beta-Binding Protein 1 (Ltbp1) Elisa Kit for detecting Latent Transforming Growth Factor Beta Binding Protein 1 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q14766; LTBP1. Target Synonyms: 9430031G15Rik; 9830146M04; Latent transforming growth factor beta binding protein 1; Latent-transforming growth factor beta-binding protein 1; Latent-transforming growth factor beta-binding protein; isoform 1L; Latent-transforming growth factor beta-binding protein; isoform 1S; LBT1S; LTBP-1; Ltbp1; LTBP1_HUMAN; MGC163161; TGF-beta1-BP-1; Tgfb; Transforming growth factor beta-1-binding protein 1; Transforming growth factor beta-1-masking protein large subunit. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4090