Learn More
Biomatik Corporation Human Lim And Sh3 Domain Protein 1 (Lasp1) Elisa Kit
Supplier:  Biomatik Corporation EKC34431
Encompass_Preferred
Human Lim And Sh3 Domain Protein 1 (Lasp1) Elisa Kit for detecting Lim And Sh3 Protein 1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 25-1600pg/ml. Sensitivity: 6.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q14847; LASP1. Target Synonyms: LASP 1; LASP-1; LASP1; LASP1_HUMAN; LIM and SH3 domain protein 1; LIM and SH3 protein 1; Metastatic lymph node gene 50 protein; MLN 50; MLN50; OTTHUMP00000164238; OTTHUMP00000164239. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4097