Learn More
Biomatik Corporation Human Lipolysis-Stimulated Lipoprotein Receptor (Lsr) Elisa Kit
Supplier:  Biomatik Corporation EKC34446
Encompass_Preferred
Human Lipolysis-Stimulated Lipoprotein Receptor (Lsr) Elisa Kit for detecting Lipolysis Stimulated Lipoprotein Receptor in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 6.25-400pg/ml. Sensitivity: 1.56pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q86X29; LSR. Target Synonyms: ILDR3; Immunoglobulin like domain containing receptor 3; Lipolysis Stimulated Lipoprotein Receptor; Lipolysis stimulated remnant; Lipolysis-stimulated lipoprotein receptor; LISCH; LISCH protein; LISCH7; Liver specific bHLH Zip transcription factor; LSR; LSR_HUMAN; MGC10659; MGC48312; MGC48503. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4099