Learn More
Biomatik Corporation Human Lysine-Specific Demethylase 5A (Kdm5A) Elisa Kit
Supplier:  Biomatik Corporation EKC34495
Encompass_Preferred
Human Lysine-Specific Demethylase 5A (Kdm5A) Elisa Kit for detecting Lysine (K)-Specific Demethylase 5A in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 23.5-1500pg/ml. Sensitivity: 5.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P29375; KDM5A. Target Synonyms: Histone demethylase JARID1A; JARID1A; Jumonji/ARID domain containing protein 1A; Jumonji/ARID domain-containing protein 1A; Kdm5a; KDM5A_HUMAN; Lysine-specific demethylase 5A; RBBP-2; RBBP2; RBP2; Retinoblastoma binding protein 2; Retinoblastoma-binding protein 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4110