Learn More
Biomatik Corporation Human Macrophage Inflammatory Protein-1Α, Mip-1Α Elisa Kit
Supplier:  Biomatik Corporation EKC34517
Encompass_Preferred
Human Macrophage Inflammatory Protein-1Α, Mip-1Α Elisa Kit for detecting Chemokine (C-C Motif) Ligand 3 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 39-2500pg/ml. Sensitivity: 9.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P10147; CCL3. Target Synonyms: C C motif chemokine 3; CCL 3; CCL3; CCL3_HUMAN; Chemokine (C C motif) ligand 3; Chemokine C C motif ligand 3; Chemokine ligand 3; G0/G1 switch regulatory protein 19 1; G0/G1 switch regulatory protein 19-1; G0S19 1; G0S19 1 protein; Heparin binding chemotaxis protein; L2G25B; LD78 alpha; LD78-alpha (4-69); LD78alpha; Macrophage inflammatory protein 1 alpha; Macrophage inflammatory protein 1-alpha; macrophage inflammatory protein 1a; MIP 1 alpha; MIP 1A; MIP-1-alpha; MIP-1-alpha (4-69); MIP1 alpha; MIP1A; PAT 464.1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4117