Learn More
Biomatik Corporation Human Malate Dehydrogenase 1 (Mdh1) Elisa Kit
Supplier:  Biomatik Corporation EKC34526
Encompass_Preferred
Human Malate Dehydrogenase 1 (Mdh1) Elisa Kit for detecting Malate Dehydrogenase 1, Nad (Soluble) in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 9.38-600mU/ml. Sensitivity: 2.35mU/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P40925; MDH1. Target Synonyms: cytoplasmic; Cytosolic malate dehydrogenase; Diiodophenylpyruvate reductase; Malate dehydrogenase 1; NAD (soluble); Malate dehydrogenase; Malate dehydrogenase cytoplasmic; MDH s; mdh1; MDHA; MDHC_HUMAN; MDHs; MGC:1375; MOR2; Soluble malate dehydrogenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4119