Learn More
Biomatik Corporation Human Mast Cell Carboxypeptidase Elisa Kit
Supplier:  Biomatik Corporation EKC34541
Encompass_Preferred
Human Mast Cell Carboxypeptidase Elisa Kit for detecting Carboxypeptidase A3 (Mast Cell) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P15088; CPA3. Target Synonyms: carboxypeptidase A; carboxypeptidase A3 (mast cell); Carboxypeptidase A3; carboxypeptidase A3 mast cell; CBPA3_HUMAN; Cpa3; Mast cell carboxypeptidase A; mast cell carboxypeptidase A3; MC CPA; MC-CPA; tissue carboxypeptidase A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4122