Learn More
Biomatik Corporation Human Mhc Class I Polypeptide-Related Sequence B (Micb) Elisa Kit
Supplier:  Biomatik Corporation EKC34607
Encompass_Preferred
Human Mhc Class I Polypeptide-Related Sequence B (Micb) Elisa Kit for detecting Mhc Class I Polypeptide-Related Sequence B in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q29980; MICB. Target Synonyms: MHC class I chain related protein B; MHC class I chain related protein B2; MHC class I like molecule PERB11.2 IMX; MHC class I mic B antigen; MHC class I polypeptide related sequence B; MHC class I polypeptide-related sequence B; MIC B; MIC-B; MICB; MICB_HUMAN; PERB11.2; Stress inducible class I homolog. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4137