Learn More
Biomatik Corporation Human Monoglyceride Lipase (Mgll) Elisa Kit
Supplier:  Biomatik Corporation EKC34643
Encompass_Preferred
Human Monoglyceride Lipase (Mgll) Elisa Kit for detecting Monoglyceride Lipase in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 39.06-2500pg/ml. Sensitivity: 9.76pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q99685; MGLL. Target Synonyms: EC 3.1.1.23; HU K5; HU-K5; HUK5; Lysophospholipase homolog; Lysophospholipase like; Lysophospholipase-like; MAGL; MGL; MGLL; MGLL_HUMAN; Monoacylglycerol lipase; Monoglyceride lipase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4146