Learn More
Biomatik Corporation Human Mucin-15 (Muc15) Elisa Kit
Supplier:  Biomatik Corporation EKC34654
Encompass_Preferred
Human Mucin-15 (Muc15) Elisa Kit for detecting Mucin 15, Cell Surface Associated in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 125-8000pg/ml. Sensitivity: 31.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q8N387; MUC15. Target Synonyms: Component II; Glycoprotein 4; Glycoprotein C; MUC 15; MUC-15; Muc15; MUC15_HUMAN; Mucin 15; Mucin 15 cell surface associated; Mucin-15; PAS3; PASIII. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4148