Learn More
Biomatik Corporation Human Myeloid Differentiation Factor 88, Myd88 Elisa Kit
Supplier:  Biomatik Corporation EKC34699
Encompass_Preferred
Human Myeloid Differentiation Factor 88, Myd88 Elisa Kit for detecting Myeloid Differentiation Primary Response Gene (88) in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q99836; MYD88. Target Synonyms: Mutant myeloid differentiation primary response 88; MYD 88; Myd88; MYD88_HUMAN; MYD88D; Myeloid differentiation marker 88; Myeloid differentiation primary response 88; Myeloid differentiation primary response gene (88); Myeloid differentiation primary response gene 88; Myeloid differentiation primary response gene; Myeloid differentiation primary response protein MyD88; OTTHUMP00000161718; OTTHUMP00000208595; OTTHUMP00000209058; OTTHUMP00000209059; OTTHUMP00000209060. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4153