Learn More
Biomatik Corporation Human N-Acetyl-Ser-Asp-Lys-Pro, Acsdkp Elisa Kit
Supplier:  Biomatik Corporation EKC34721
Encompass_Preferred
Human N-Acetyl-Ser-Asp-Lys-Pro, Acsdkp Elisa Kit for detecting N-Acetyl-Ser-Asp-Lys-Pro, Acsdkp in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.235-15ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4156