Learn More
Biomatik Corporation Human Nad-Dependent Deacetylase Sirtuin-3, Mitochondrial (Sirt3) Elisa Kit
Supplier:  Biomatik Corporation EKC34729
Encompass_Preferred
Human Nad-Dependent Deacetylase Sirtuin-3, Mitochondrial (Sirt3) Elisa Kit for detecting Sirtuin (Silent Mating Type Information Regulation 2 Homolog) 3 (S. Cerevisiae) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9NTG7; SIRT3. Target Synonyms: hSIRT 3; hSIRT3; Mitochondrial nicotinamide adenine dinucleotide dependent deacetylase; NAD dependent deacetylase sirtuin 3 mitochondrial; NAD-dependent protein deacetylase sirtuin-3; mitochondrial; Regulatory protein SIR2 homolog 3; Silent mating type information regulation 2 S.cerevisiae homolog 3; Sir 2 like 3; SIR 2 like protein 3; SIR 3; SIR2 L3; Sir2 like 3; SIR2 like protein 3; SIR2-like protein 3; SIR2L3; SIR3_HUMAN; SIRT 3; SIRT3; Sirtuin 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4158